Assay Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009148.3 | NP_033174.2 | 3 - 4 | 475 | 98 | |
| GenBank mRNA | AF022962.1 | - | 3 - 4 | 501 | 98 | |
| AK010695.1 | - | 3 - 4 | 490 | 98 | ||
| AK014499.1 | - | 3 - 4 | 498 | 98 | ||
| AK028250.1 | - | 1 - 2 | 208 | 98 | ||
| AK036507.1 | - | 3 - 4 | 487 | 98 | ||
| AK038125.1 | - | 3 - 4 | 498 | 98 | ||
| AK039983.1 | - | 3 - 4 | 498 | 98 | ||
| AK086603.1 | - | 3 - 4 | 494 | 98 | ||
| AK173235.1 | - | 3 - 4 | 478 | 98 | ||
| BC034644.1 | - | 3 - 4 | 475 | 5355276 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028250.1 transcripts.
Set Membership: |
|

Exoc4
Genomic Map