Assay Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009148.3 | NP_033174.2 | 4 - 5 | 661 | 65 | |
| GenBank mRNA | AF022962.1 | - | 4 - 5 | 687 | 65 | |
| AK010695.1 | - | 4 - 5 | 676 | 65 | ||
| AK014499.1 | - | 4 - 5 | 684 | 65 | ||
| AK028250.1 | - | 2 - 3 | 394 | 65 | ||
| AK036507.1 | - | 4 - 5 | 673 | 65 | ||
| AK038125.1 | - | 4 - 5 | 684 | 65 | ||
| AK039983.1 | - | 4 - 5 | 684 | 65 | ||
| AK086603.1 | - | 4 - 5 | 680 | 65 | ||
| AK173235.1 | - | 4 - 5 | 664 | 65 | ||
| AK199139.1 | - | 1 - 2 | 40 | 65 | ||
| AK212838.1 | - | 1 - 2 | 40 | 65 | ||
| BC034644.1 | - | 4 - 5 | 661 | 5355276 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028250.1 transcripts.
Set Membership: |
|

Exoc4
Genomic Map