Assay Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009148.3 | NP_033174.2 | 11 - 12 | 1745 | 65 | |
| GenBank mRNA | AF022962.1 | - | 11 - 12 | 1771 | 65 | |
| AK028250.1 | - | 9 - 10 | 1477 | 65 | ||
| AK039983.1 | - | 11 - 12 | 1768 | 65 | ||
| AK173235.1 | - | 11 - 12 | 1748 | 65 | ||
| BC034644.1 | - | 11 - 12 | 1745 | 5355276 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc4
Genomic Map