Assay Details
| Gene Symbol: Sftpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009160.2 | NP_033186.1 | 5 - 6 | 626 | 114 | |
| GenBank mRNA | BC003705.1 | - | 5 - 6 | 577 | 3601620 | 114 |
| BC012896.1 | - | 4 - 5 | 814 | 114 | ||
| BG085186.2 | - | 1 - 2 | 65 | 114 | ||
| L40156.1 | - | 4 - 5 | 554 | 114 | ||
More Information
Set Membership: |
|

Sftpd
Genomic Map