Assay Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009203.3 | NP_033229.3 | 9 - 10 | 1918 | 66 | |
| GenBank mRNA | AB005451.1 | - | 9 - 10 | 1699 | 66 | |
| AI314749.1 | - | 1 - 2 | 66 | |||
| AK142580.1 | - | 8 - 9 | 1067 | 66 | ||
| BC035927.1 | - | 1854 | 4242354 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036236 | AI314749.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036236 | AK142580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036236 | BC035927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036236 | NM_009203.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22031 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149078 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036236 | AI314749.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036236 | AK142580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036236 | BC035927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036236 | NM_009203.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335519 | AB005451.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335519 | BC035927.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335519 | NM_009203.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31935497 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149078 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09096739 | - | - |
| Whole Mouse Genome G4122A | A_51_P335519 | AB005451.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335519 | BC035927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335519 | NM_009203.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113930 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267040 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267035 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14267052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31935540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14407399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14371133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14267043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14482482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14482483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14343255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14407391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14343259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14371129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14371132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14143420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14420010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14343256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14479246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14479240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14267044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14150578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31935477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465414 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465421 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422898_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422897_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465411 | - | - |
| Mouse Genome 430 2.0 Array | 1422897_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422897_at | - | - |
| Mouse Genome 430 2.0 Array | 1422898_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1422898_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20521_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20521_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900009358.mm.1 | NM_009203.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009358.mm.1 | NM_009203.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009358.mm.1 | TC1900001012.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009357.mm.1 | NM_009203.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009357.mm.1 | NM_009203.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009357.mm.1 | TC1900001012.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422897_PM_at | - | - |
| Mouse Expression Set 430 | 1422897_at | - | - |
| Mouse Expression Set 430 | 1422898_PM_s_at | - | - |
| Mouse Expression Set 430 | 1422898_s_at | - | - |
| Murine Genome U74v2 Set | 102429_at | - | - |
| Murine Genome U74v2 Set | 102429_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422897_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422898_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422898_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422897_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422897_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422898_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422897_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422898_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4230253 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4920288 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4230253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255257 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251401 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255257 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251401 | - | - |
| mouse-6 v1.1 (Illumina) | scl53539.9.129_6-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000442.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920288 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53539.9.129_6-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000442.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251401 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251401 | - | - |

Slc22a12
Genomic Map