Assay Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009205.2 | NP_033231.2 | 6 - 7 | 1222 | 87 | |
| GenBank mRNA | AK052623.1 | - | 6 - 7 | 1193 | 87 | |
| AK132275.1 | - | 6 - 7 | 1194 | 87 | ||
| BC013441.1 | - | 6 - 7 | 1164 | 4222972 | 87 | |
| D88533.1 | - | 6 - 7 | 1182 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc3a1
Genomic Map