Assay Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009205.2 | NP_033231.2 | 8 - 9 | 1583 | 110 | |
| GenBank mRNA | AK052623.1 | - | 8 - 9 | 1554 | 110 | |
| AK132275.1 | - | 8 - 9 | 1555 | 110 | ||
| BC013441.1 | - | 8 - 9 | 1525 | 4222972 | 110 | |
| D88533.1 | - | 8 - 9 | 1543 | 110 | ||
More Information
Set Membership: |
|

Slc3a1
Genomic Map