Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009213.2 | NP_033239.1 | 1 - 2 | 357 | 85 | |
| GenBank mRNA | AJ222800.1 | - | 1 - 2 | 358 | 85 | |
| AK075616.1 | - | 1 - 2 | 367 | 85 | ||
| BC010978.1 | - | 1 - 2 | 367 | 4193282 | 85 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ222800.1,AK075616.1,BC010978.1,NM_009213.2 transcripts.
Set Membership: |
|

Smpd2
Genomic Map