Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009213.2 | NP_033239.1 | 8 - 9 | 1037 | 62 | |
| GenBank mRNA | AJ222800.1 | - | 8 - 9 | 1038 | 62 | |
| AK075616.1 | - | 8 - 9 | 1047 | 62 | ||
| AK088962.1 | - | 5 - 6 | 1805 | 62 | ||
| AK161573.1 | - | 2149 | 62 | |||
| BC010978.1 | - | 8 - 9 | 1047 | 4193282 | 62 | |
More Information
Set Membership: |
|

Smpd2
Genomic Map