Assay Details
| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009228.2 | NP_033254.2 | 4 - 5 | 1006 | 75 | |
| GenBank mRNA | AK137379.1 | - | 4 - 5 | 1007 | 75 | |
| BC018546.1 | - | 4 - 5 | 956 | 3674921 | 75 | |
| U00677.1 | - | 4 - 5 | 1003 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snta1
Genomic Map