Assay Details
| Gene Symbol: Sntb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009229.4 | NP_033255.1 | 1 - 2 | 576 | 113 | |
| GenBank mRNA | AK040423.1 | - | 1 - 2 | 577 | 113 | |
| AK080059.1 | - | 1 - 2 | 573 | 113 | ||
| AK131687.1 | - | 1 - 2 | 601 | 113 | ||
| AK165009.1 | - | 1 - 2 | 578 | 113 | ||
| AK165900.1 | - | 1 - 2 | 553 | 113 | ||
| BC030352.1 | - | 6 - 7 | 113 | |||
| BC038704.1 | - | 6 - 7 | 113 | |||
| BC051571.1 | - | 4 - 5 | 113 | |||
| BC138850.1 | - | 1 - 2 | 575 | 8861872 | 113 | |
| BC145442.1 | - | 1 - 2 | 575 | 9054000 | 113 | |
| U00678.1 | - | 1 - 2 | 575 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sntb2
Genomic Map