Assay Details
| Gene Symbol: Soat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009230.3 | NP_033256.2 | 6 - 7 | 684 | 103 | |
| GenBank mRNA | AK052299.1 | - | 3 - 4 | 241 | 103 | |
| AK054149.1 | - | 6 - 7 | 597 | 103 | ||
| AK142486.1 | - | 6 - 7 | 601 | 103 | ||
| AK159393.1 | - | 6 - 7 | 614 | 103 | ||
| AK159529.1 | - | 6 - 7 | 614 | 103 | ||
| AK160017.1 | - | 6 - 7 | 614 | 103 | ||
| BC083092.1 | - | 5 - 6 | 498 | 30446741 | 103 | |
| DQ903181.1 | - | 6 - 7 | 601 | 103 | ||
| S81092.1 | - | 6 - 7 | 618 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Soat1
Genomic Map