Assay Details
| Gene Symbol: Stim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009287.4 | NP_033313.2 | 3 - 4 | 981 | 66 | |
| GenBank mRNA | AK041944.1 | - | 3 - 4 | 1000 | 66 | |
| AK201690.1 | - | 3 - 4 | 282 | 66 | ||
| BC021644.1 | - | 3 - 4 | 983 | 4191156 | 66 | |
| U47323.1 | - | 3 - 4 | 583 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stim1
Genomic Map