Assay Details
| Gene Symbol: S100g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009789.2 | NP_033919.1 | 2 - 3 | 231 | 93 | |
| GenBank mRNA | AF028071.1 | - | 2 - 3 | 197 | 93 | |
| AF136283.1 | - | 2 - 3 | 194 | 93 | ||
| AF293948.1 | - | 2 - 3 | 231 | 93 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013236 | NM_009789.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013236 | AF293948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013236 | AF293949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111163 | AK146558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111163 | BC010751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111163 | NM_009789.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111163 | AF136283.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111163 | AF293948.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151373 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013236 | NM_009789.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013236 | AF293948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013236 | AF293949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111163 | AK146558.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111163 | BC010751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111163 | NM_009789.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111163 | AF136283.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111163 | AF293948.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32791657 | - | - |
| Whole Mouse Genome G4122A | A_51_P118545 | AK146558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118545 | BC010751.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118545 | NM_009789.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118545 | AF028071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P118545 | AF136283.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15008375 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15107226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15107227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15109869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15202469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32791682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15109868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15019873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14860737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14537501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15202467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14738799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14537504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14738796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15047297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15097255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32791653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607708 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448964_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607705 | - | - |
| Mouse Genome 430 2.0 Array | 1448964_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442998_at | - | - |
| Mouse Genome 430 2.0 Array | 1448964_at | - | - |
| Mouse Genome 430 2.0 Array | 1442998_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12309_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12309_at | - | - |
| Mouse Expression Set 430 | 1448964_PM_at | - | - |
| Mouse Expression Set 430 | 1448964_at | - | - |
| Murine Genome U74v2 Set | 160918_PM_at | - | - |
| Murine Genome U74v2 Set | 161835_PM_at | - | - |
| Murine Genome U74v2 Set | 160918_at | - | - |
| Murine Genome U74v2 Set | 161835_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448964_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448964_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448964_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448964_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 531763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2340064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249485 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249485 | - | - |
| mouse-6 v1.1 (Illumina) | scl53622.3.4_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53622.3.4_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1249485 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249485 | - | - |

S100g
Genomic Map