Assay Details
| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009814.3 | NP_033944.2 | 5 - 6 | 789 | 88 | |
| GenBank mRNA | AF068244.1 | - | 5 - 6 | 734 | 88 | |
| BC092229.1 | - | 5 - 6 | 702 | 30603069 | 88 | |
| BC137668.1 | - | 5 - 6 | 698 | 8860687 | 88 | |
| BC140959.1 | - | 5 - 6 | 698 | 9055571 | 88 | |
| U91483.1 | - | 5 - 6 | 608 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casq2
Genomic Map