Assay Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009840.3 | NP_033970.3 | 9 - 10 | 1091 | 83 | |
| GenBank mRNA | AK010882.1 | - | 9 - 10 | 1092 | 83 | |
| AK014005.1 | - | 2 - 3 | 157 | 83 | ||
| AK075711.1 | - | 9 - 10 | 1072 | 83 | ||
| AK075880.1 | - | 9 - 10 | 1080 | 83 | ||
| AK145661.1 | - | 9 - 10 | 1075 | 83 | ||
| AK145754.1 | - | 9 - 10 | 1074 | 83 | ||
| AK145918.1 | - | 9 - 10 | 1093 | 83 | ||
| AK147038.1 | - | 9 - 10 | 1094 | 83 | ||
| AK159969.1 | - | 9 - 10 | 1083 | 83 | ||
| AK168465.1 | - | 9 - 10 | 1064 | 83 | ||
| AK172867.1 | - | 9 - 10 | 1062 | 83 | ||
| AK218945.1 | - | 2 - 3 | 83 | |||
| BC009007.1 | - | 9 - 10 | 1049 | 3582639 | 83 | |
| Z37164.1 | - | 9 - 10 | 1095 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct8
Genomic Map