Assay Details
| Gene Symbol: Cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009840.3 | NP_033970.3 | 11 - 12 | 1288 | 76 | |
| GenBank mRNA | AK010882.1 | - | 11 - 12 | 1289 | 76 | |
| AK014005.1 | - | 4 - 5 | 354 | 76 | ||
| AK075711.1 | - | 11 - 12 | 1269 | 76 | ||
| AK075880.1 | - | 11 - 12 | 1277 | 76 | ||
| AK145661.1 | - | 11 - 12 | 1272 | 76 | ||
| AK145754.1 | - | 11 - 12 | 1271 | 76 | ||
| AK145918.1 | - | 11 - 12 | 1290 | 76 | ||
| AK147038.1 | - | 11 - 12 | 1291 | 76 | ||
| AK159969.1 | - | 11 - 12 | 1279 | 76 | ||
| AK168465.1 | - | 11 - 12 | 1261 | 76 | ||
| AK172867.1 | - | 11 - 12 | 1259 | 76 | ||
| BC009007.1 | - | 11 - 12 | 1246 | 3582639 | 76 | |
| Z37164.1 | - | 11 - 12 | 1292 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct8
Genomic Map