Assay Details
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161623.1 | NP_001155095.1 | 9 - 10 | 764 | 60 | |
| NM_009862.2 | NP_033992.2 | 11 - 12 | 1141 | 60 | ||
| GenBank mRNA | AF081536.1 | - | 11 - 12 | 1141 | 60 | |
| AF081537.1 | - | 10 - 11 | 840 | 60 | ||
| AF081538.1 | - | 10 - 11 | 840 | 60 | ||
| AF098068.1 | - | 10 - 11 | 887 | 60 | ||
| AJ223729.1 | - | 10 - 11 | 861 | 60 | ||
| AK147013.1 | - | 10 - 11 | 858 | 60 | ||
| AK167850.1 | - | 9 - 10 | 752 | 60 | ||
| BC006856.1 | - | 10 - 11 | 856 | 60 | ||
| BC028635.1 | - | 10 - 11 | 894 | 1262927 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc45
Genomic Map