Assay Details
| Gene Symbol: Cdc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161623.1 | NP_001155095.1 | 16 - 17 | 1570 | 64 | |
| NM_009862.2 | NP_033992.2 | 18 - 19 | 1947 | 64 | ||
| GenBank mRNA | AF081536.1 | - | 18 - 19 | 1947 | 64 | |
| AF081537.1 | - | 17 - 18 | 1646 | 64 | ||
| AF081538.1 | - | 17 - 18 | 1646 | 64 | ||
| AF081539.1 | - | 5 - 6 | 531 | 64 | ||
| AF098068.1 | - | 17 - 18 | 1693 | 64 | ||
| AJ223729.1 | - | 17 - 18 | 1667 | 64 | ||
| AK147013.1 | - | 17 - 18 | 1664 | 64 | ||
| AK167850.1 | - | 16 - 17 | 1558 | 64 | ||
| BC006856.1 | - | 17 - 18 | 1662 | 64 | ||
| BC028635.1 | - | 17 - 18 | 1700 | 1262927 | 64 | |
| CO040149.1 | - | 2 - 3 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc45
Genomic Map