Assay Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009906.5 | NP_034036.1 | 5 - 6 | 530 | 73 | |
| GenBank mRNA | AF111172.1 | - | 5 - 6 | 538 | 73 | |
| AJ011912.1 | - | 5 - 6 | 524 | 73 | ||
| AK002418.1 | - | 5 - 6 | 549 | 73 | ||
| AK048279.1 | - | 5 - 6 | 531 | 73 | ||
| AK083832.1 | - | 4 - 5 | 1795 | 73 | ||
| AK146759.1 | - | 5 - 6 | 523 | 73 | ||
| AK155021.1 | - | 5 - 6 | 549 | 73 | ||
| AK165669.1 | - | 5 - 6 | 555 | 73 | ||
| AK166648.1 | - | 3 - 4 | 324 | 73 | ||
| AK169924.1 | - | 5 - 6 | 537 | 73 | ||
| AK170781.1 | - | 5 - 6 | 526 | 73 | ||
| BC024820.1 | - | 5 - 6 | 532 | 5360558 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083832.1 transcripts.
Set Membership: |
|

Tpp1
Genomic Map