Assay Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009949.2 | NP_034079.2 | 3 - 4 | 469 | 72 | |
| GenBank mRNA | AK144441.1 | - | 3 - 4 | 469 | 72 | |
| AK150163.1 | - | 3 - 4 | 441 | 72 | ||
| AK169038.1 | - | 3 - 4 | 366 | 72 | ||
| BC138514.1 | - | 3 - 4 | 404 | 8861534 | 72 | |
| BC145859.1 | - | 3 - 4 | 404 | 40131162 | 72 | |
| CD545813.1 | - | 3 - 4 | 444 | 72 | ||
| U01163.1 | - | 3 - 4 | 414 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpt2
Genomic Map