Assay Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010176.4 | NP_034306.2 | 9 - 10 | 1058 | 72 | |
| GenBank mRNA | AK143759.1 | - | 9 - 10 | 887 | 72 | |
| AK158808.1 | - | 9 - 10 | 944 | 72 | ||
| BC010767.1 | - | 9 - 10 | 843 | 4242421 | 72 | |
| M84145.1 | - | 9 - 10 | 884 | 72 | ||
| Z11774.1 | - | 9 - 10 | 870 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fah
Genomic Map