Assay Details
| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010221.2 | NP_034351.2 | 6 - 7 | 1148 | 113 | |
| GenBank mRNA | AK019446.2 | - | 6 - 7 | 1087 | 113 | |
| BC029546.1 | - | 6 - 7 | 1095 | 5059670 | 113 | |
| L07063.1 | - | 6 - 7 | 1151 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp10
Genomic Map