Assay Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017861.1 | - | 2 - 3 | 322 | 65 | |
| AK146459.1 | - | 3 - 4 | 438 | 65 | ||
| AK158704.1 | - | 3 - 4 | 440 | 65 | ||
| BC008130.1 | - | 4 - 5 | 443 | 3498984 | 65 | |
| BC034861.1 | - | 3 - 4 | 447 | 65 | ||
| X71978.1 | - | 3 - 4 | 462 | 65 | ||
| Z67963.1 | - | 3 - 4 | 462 | 65 | ||
More Information
Set Membership: |
|

Aktip
Genomic Map