Assay Details
| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010247.2 | NP_034377.2 | 8 - 9 | 1246 | 72 | |
| GenBank mRNA | AB010282.1 | - | 8 - 9 | 1187 | 72 | |
| AF483500.1 | - | 7 - 8 | 1126 | 72 | ||
| AF483501.1 | - | 7 - 8 | 1126 | 72 | ||
| AK088530.1 | - | 2 - 3 | 197 | 72 | ||
| AK143570.1 | - | 8 - 9 | 1221 | 72 | ||
| AK146392.1 | - | 8 - 9 | 1165 | 72 | ||
| AK146394.1 | - | 8 - 9 | 1165 | 72 | ||
| AK146396.1 | - | 8 - 9 | 1213 | 72 | ||
| BC031422.1 | - | 8 - 9 | 1195 | 3669715 | 72 | |
| BC051085.1 | - | 8 - 9 | 2424 | 72 | ||
| M38700.1 | - | 7 - 8 | 1088 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrcc6
Genomic Map