Assay Details
| Gene Symbol: Tpsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024374.1 | - | 1 - 2 | 83 | 4983727 | 87 |
| L31853.1 | - | 1 - 2 | 77 | 87 | ||
| M57626.1 | - | 2 - 3 | 105 | 87 | ||
| X78542.1 | - | 77 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsb2
Genomic Map