Assay Details
| Gene Symbol: Mettl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010792.1 | NP_034922.1 | 4 - 5 | 605 | 95 | |
| GenBank mRNA | AB023619.1 | - | 4 - 5 | 605 | 95 | |
| AK160562.1 | - | 4 - 5 | 584 | 95 | ||
| AK160914.1 | - | 4 - 5 | 582 | 95 | ||
| BC012649.1 | - | 4 - 5 | 614 | 4020487 | 95 | |
| Y18644.1 | - | 4 - 5 | 551 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533650 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533641 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533638 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533649 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533647 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533644 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533643 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065465 | AK160562.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065465 | AK160914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065465 | BC012649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065465 | Y18644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065465 | NM_010792.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794326 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159211 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065465 | AK160562.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065465 | AK160914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065465 | BC012649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065465 | Y18644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065465 | NM_010792.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499103 | AK160562.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499103 | AK160914.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499103 | AK164284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499103 | Y18644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499103 | NM_010792.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159211 | - | - |
| Whole Mouse Genome G4122A | A_51_P499103 | AK160562.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499103 | AK160914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499103 | AK164284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499103 | Y18644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499103 | NM_010792.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140555 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08904536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08904523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08904518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08904522 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08904526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533647 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533644 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533638 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533641 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533649 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533650 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533643 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27533645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08737291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08415643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08290919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08925825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08290918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08904522 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10366798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366802 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447683_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419122_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439155_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10366796 | - | - |
| Mouse Genome 430 2.0 Array | 1439155_at | - | - |
| Mouse Genome 430 2.0 Array | 1419122_at | - | - |
| Mouse Genome 430 2.0 Array | 1439155_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447683_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419122_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447683_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17299_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17299_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000010917.mm.1 | NM_010792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000010917.mm.1 | NM_010792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000010917.mm.1 | TC1000001544.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000010916.mm.1 | NM_010792.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000010916.mm.1 | NM_010792.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000010916.mm.1 | TC1000001544.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419122_at | - | - |
| Mouse Expression Set 430 | 1419122_PM_at | - | - |
| Mouse Expression Set 430 | 1439155_at | - | - |
| Mouse Expression Set 430 | 1447683_PM_x_at | - | - |
| Mouse Expression Set 430 | 1447683_x_at | - | - |
| Mouse Expression Set 430 | 1439155_PM_at | - | - |
| Murine Genome U74v2 Set | 99499_PM_at | - | - |
| Murine Genome U74v2 Set | 99499_at | - | - |
| Murine Genome U74v2 Set | 167076_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167076_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419122_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419122_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419122_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419122_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439155_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439155_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447683_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447683_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381433 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7040433 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6180270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4540670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6180270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7040433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732419 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240222 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2612541 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240222 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732419 | - | - |
| mouse-6 v1.1 (Illumina) | scl38335.7.1_58-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003887.1_47-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003890.1_115-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180270 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040433 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003890.1_115-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003887.1_47-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38335.7.1_58-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540670 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3001946 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240222 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732419 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732419 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240222 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3001946 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003890.1_115-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38335.7.1_58-S | - | - |

Mettl1
Genomic Map