Assay Details
| Gene Symbol: Rad9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011237.2 | NP_035367.1 | 1 - 2 | 126 | 119 | |
| GenBank mRNA | AF045663.1 | - | 1 - 2 | 126 | 119 | |
| AK134293.1 | - | 1 - 2 | 139 | 119 | ||
| AK138973.1 | - | 1 - 2 | 131 | 119 | ||
| AK165833.1 | - | 1 - 2 | 161 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF045663.1, AK134293.1, NM_011237.2 transcripts.
Set Membership: |
|

Rad9a
Genomic Map