Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 14 - 15 | 1496 | 72 | |
| NM_001271408.1 | NP_001258337.1 | 14 - 15 | 1380 | 72 | ||
| NM_001271409.1 | NP_001258338.1 | 3 - 4 | 361 | 72 | ||
| NM_011366.3 | NP_035496.1 | 14 - 15 | 1383 | 72 | ||
| GenBank mRNA | AB190911.1 | - | 14 - 15 | 1497 | 72 | |
| AF064806.1 | - | 14 - 15 | 1349 | 72 | ||
| AK029703.1 | - | 16 - 17 | 1494 | 72 | ||
| AK164607.1 | - | 2 - 3 | 1825 | 72 | ||
| AK165689.1 | - | 14 - 15 | 1390 | 72 | ||
| BC030933.1 | - | 13 - 14 | 1197 | 4920799 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map