Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 19 - 20 | 2193 | 81 | |
| NM_001271408.1 | NP_001258337.1 | 19 - 20 | 2077 | 81 | ||
| NM_001271409.1 | NP_001258338.1 | 8 - 9 | 1058 | 81 | ||
| NM_011366.3 | NP_035496.1 | 19 - 20 | 2080 | 81 | ||
| GenBank mRNA | AB190911.1 | - | 19 - 20 | 2194 | 81 | |
| AF064806.1 | - | 19 - 20 | 2046 | 81 | ||
| AK029703.1 | - | 21 - 22 | 2192 | 81 | ||
| AK165689.1 | - | 19 - 20 | 2087 | 81 | ||
| BC030933.1 | - | 18 - 19 | 1894 | 4920799 | 81 | |
| U58889.1 | - | 2 - 3 | 362 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map