Assay Details
| Gene Symbol: St6galnac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276425.1 | NP_001263354.1 | 4 - 5 | 851 | 76 | |
| NM_011373.3 | NP_035503.1 | 3 - 4 | 1204 | 76 | ||
| GenBank mRNA | AJ007310.1 | - | 4 - 5 | 846 | 76 | |
| AK085730.1 | - | 4 - 5 | 842 | 76 | ||
| AK144884.1 | - | 4 - 5 | 842 | 76 | ||
| AK153036.1 | - | 4 - 5 | 811 | 76 | ||
| AK159712.1 | - | 3 - 5 | 1195 | 76 | ||
| AK161559.1 | - | 4 - 5 | 836 | 76 | ||
| BC056451.1 | - | 3 - 4 | 657 | 5685866 | 76 | |
| BC086784.1 | - | 792 | 6844232 | 76 | ||
| Y15779.1 | - | 4 - 5 | 828 | 76 | ||
| Y15780.1 | - | 3 - 4 | 840 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St6galnac4
Genomic Map