Assay Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113559.2 | NP_001107031.2 | 13 - 14 | 2043 | 63 | |
| NM_001243163.1 | NP_001230092.1 | 13 - 14 | 1935 | 63 | ||
| NM_011444.3 | NP_035574.2 | 14 - 15 | 2187 | 63 | ||
| GenBank mRNA | AB006330.1 | - | 14 - 15 | 2099 | 63 | |
| AJ010604.1 | - | 13 - 14 | 1925 | 63 | ||
| AK160154.1 | - | 7 - 8 | 1001 | 63 | ||
| BC110478.2 | - | 13 - 14 | 1927 | 40047865 | 63 | |
| KF793916.1 | - | 14 - 15 | 1888 | 63 | ||
More Information
Set Membership: |
|

Sox5
Genomic Map