Assay Details
| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025559.3 | NP_001020730.1 | 13 - 14 | 1753 | 67 | |
| NM_001025560.2 | NP_001020731.1 | 13 - 14 | 1867 | 67 | ||
| NM_001277326.1 | NP_001264255.1 | 13 - 14 | 1848 | 67 | ||
| NM_001277327.1 | NP_001264256.1 | 12 - 13 | 1908 | 67 | ||
| NM_001277328.1 | NP_001264257.1 | 11 - 12 | 1613 | 67 | ||
| NM_011445.4 | NP_035575.1 | 14 - 15 | 1901 | 67 | ||
| GenBank mRNA | AJ010605.1 | - | 2 - 3 | 180 | 67 | |
| AK029696.1 | - | 11 - 12 | 1541 | 67 | ||
| AK030713.1 | - | 13 - 14 | 1868 | 67 | ||
| AK132409.1 | - | 13 - 14 | 1814 | 67 | ||
| BC067407.1 | - | 12 - 13 | 1905 | 30094413 | 67 | |
| D61689.1 | - | 11 - 12 | 1609 | 67 | ||
| U32614.1 | - | 14 - 15 | 1905 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox6
Genomic Map