Assay Details
| Gene Symbol: Racgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253808.1 | NP_001240737.1 | 6 - 7 | 710 | 85 | |
| NM_001253809.1 | NP_001240738.1 | 6 - 7 | 706 | 85 | ||
| NM_012025.7 | NP_036155.1 | 7 - 8 | 841 | 85 | ||
| GenBank mRNA | AB030252.1 | - | 6 - 7 | 690 | 85 | |
| AF079974.1 | - | 6 - 7 | 651 | 85 | ||
| AF212320.1 | - | 6 - 7 | 677 | 85 | ||
| AF212321.1 | - | 7 - 8 | 808 | 85 | ||
| AK053850.1 | - | 6 - 7 | 671 | 85 | ||
| AK144608.1 | - | 7 - 8 | 809 | 85 | ||
| AK144745.1 | - | 6 - 7 | 670 | 85 | ||
| AK154929.1 | - | 7 - 8 | 829 | 85 | ||
| AK162660.1 | - | 4 - 5 | 3910 | 85 | ||
| AK165897.1 | - | 6 - 7 | 663 | 85 | ||
| AK166084.1 | - | 7 - 8 | 799 | 85 | ||
| AK166094.1 | - | 6 - 7 | 668 | 85 | ||
| AK168020.1 | - | 6 - 7 | 667 | 85 | ||
| AK168025.1 | - | 6 - 7 | 667 | 85 | ||
| AK168170.1 | - | 6 - 7 | 667 | 85 | ||
| AK168679.1 | - | 6 - 7 | 651 | 85 | ||
| AK181912.1 | - | 3 - 4 | 235 | 85 | ||
| AK183940.1 | - | 4 - 5 | 301 | 85 | ||
| AK184356.1 | - | 2 - 3 | 140 | 85 | ||
| AK200143.1 | - | 2 - 3 | 140 | 85 | ||
| AK207314.1 | - | 4 - 5 | 301 | 85 | ||
| BC010715.1 | - | 6 - 7 | 656 | 3602242 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Racgap1
Genomic Map