Assay Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013606.1 | NP_038634.1 | 2 - 3 | 208 | 90 | |
| NR_003508.1 | 2 - 3 | 210 | 90 | |||
| GenBank mRNA | AB029920.1 | - | 2 - 3 | 208 | 90 | |
| AK143831.1 | - | 2 - 3 | 211 | 90 | ||
| AK147949.1 | - | 830 | 90 | |||
| BC007127.1 | - | 2 - 3 | 193 | 3591798 | 90 | |
| J03368.1 | - | 2 - 3 | 208 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 797835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125973 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179748 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152131 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | J03368.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | J03368.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30999450 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03267083 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179748 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08698258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03267069 | - | - |
| Whole Mouse Genome G4122A | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | BC007127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | J03368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | AK143831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | NR_003508.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P514085 | AB029920.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112478 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179748 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152131 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853493 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853498 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12853491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30999621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12924674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12977129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12576616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12684651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12853502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12924670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12576617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12924657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12981607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12853498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12853504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30999454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437231 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419676_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437224 | - | - |
| Mouse Genome 430 2.0 Array | 1419676_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419676_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17858_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17858_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600008636.mm.1 | NR_003508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008636.mm.1 | NR_003508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008636.mm.1 | TC1600001092.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008634.mm.1 | NR_003508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008634.mm.1 | NR_003508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008634.mm.1 | TC1600001092.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600008635.mm.1 | NR_003508.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008635.mm.1 | NR_003508.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008635.mm.1 | TC1600001092.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419676_at | - | - |
| Mouse Expression Set 430 | 1419676_PM_at | - | - |
| Murine Genome U74v2 Set | 102699_PM_at | - | - |
| Murine Genome U74v2 Set | 102699_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419676_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419676_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419676_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419676_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 837469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2030373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667746 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2667746 | - | - |
| mouse-6 v1.1 (Illumina) | scl017858.14_123-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017858.14_123-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239219 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239219 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017858.14_123-S | - | - |

Mx2
Genomic Map