Assay Details
| Gene Symbol: Syn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164495.1 | NP_001157967.1 | 3 - 4 | 624 | 81 | |
| NM_013722.3 | NP_038750.2 | 3 - 4 | 624 | 81 | ||
| GenBank mRNA | AF192754.1 | - | 3 - 4 | 569 | 81 | |
| AF498252.1 | - | 2 - 3 | 371 | 81 | ||
| AF498253.1 | - | 2 - 3 | 371 | 81 | ||
| AK043854.1 | - | 3 - 4 | 625 | 81 | ||
| AK048643.1 | - | 3 - 4 | 662 | 81 | ||
| AK080557.1 | - | 3 - 4 | 585 | 81 | ||
| AK082445.1 | - | 3 - 4 | 662 | 81 | ||
| AK084032.1 | - | 3 - 4 | 610 | 81 | ||
| AK086955.1 | - | 571 | 81 | |||
| AK087342.1 | - | 4 - 5 | 700 | 81 | ||
| BC107397.2 | - | 2 - 3 | 490 | 40058225 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syn3
Genomic Map