Assay Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016685.2 | NP_057894.2 | 7 - 8 | 135 | ||
| GenBank mRNA | AF033530.1 | - | 7 - 8 | 135 | ||
| BC128368.1 | - | 7 - 8 | 769 | 40111643 | 135 | |
| BC128369.1 | - | 7 - 8 | 768 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Comp
Genomic Map