Assay Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016685.2 | NP_057894.2 | 15 - 16 | 1733 | 82 | |
| GenBank mRNA | AF033530.1 | - | 15 - 16 | 1723 | 82 | |
| BC128368.1 | - | 15 - 16 | 1723 | 40111643 | 82 | |
| BC128369.1 | - | 15 - 16 | 1722 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Comp
Genomic Map