Assay Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016795.3 | NP_058075.2 | 7 - 8 | 627 | 73 | |
| GenBank mRNA | AB012290.1 | - | 6 - 7 | 585 | 73 | |
| AJ224115.1 | - | 7 - 8 | 621 | 73 | ||
| AK015754.1 | - | 4 - 5 | 369 | 73 | ||
| AK017187.1 | - | 7 - 8 | 624 | 73 | ||
| AK140761.1 | - | 37 | 73 | |||
| AK141271.1 | - | 7 - 8 | 694 | 73 | ||
| AK151152.1 | - | 7 - 8 | 690 | 73 | ||
| AK161346.1 | - | 7 - 8 | 605 | 73 | ||
| AK161388.1 | - | 7 - 8 | 606 | 73 | ||
| BC005707.1 | - | 7 - 8 | 596 | 3157835 | 73 | |
| BC050761.1 | - | 7 - 8 | 627 | 6771151 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk1
Genomic Map