Assay Details
| Gene Symbol: Mrpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017404.4 | NP_059100.3 | 5 - 6 | 669 | 128 | |
| GenBank mRNA | AF239728.1 | - | 5 - 6 | 586 | 128 | |
| AK007681.1 | - | 5 - 6 | 605 | 128 | ||
| AK031942.1 | - | 5 - 6 | 590 | 128 | ||
| BC012274.1 | - | 5 - 6 | 548 | 3981658 | 128 | |
| BC016561.1 | - | 5 - 6 | 605 | 2645316 | 128 | |
| BC036276.1 | - | 5 - 6 | 605 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl39
Genomic Map