Assay Details
| Gene Symbol: Smarcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018817.2 | NP_061287.2 | 9 - 10 | 1833 | 59 | |
| GenBank mRNA | AF088884.1 | - | 9 - 10 | 1813 | 59 | |
| AK077878.1 | - | 9 - 10 | 1811 | 59 | ||
| AK083488.1 | - | 9 - 10 | 1811 | 59 | ||
| AK146767.1 | - | 9 - 10 | 1828 | 59 | ||
| AK161570.1 | - | 9 - 10 | 1753 | 59 | ||
| BC029078.1 | - | 9 - 10 | 1768 | 3992325 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcal1
Genomic Map