Assay Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166531.1 | NP_001160003.1 | 4 - 5 | 397 | 79 | |
| NM_001166532.1 | NP_001160004.1 | 3 - 4 | 491 | 79 | ||
| NM_019460.2 | NP_062333.1 | 3 - 4 | 348 | 79 | ||
| GenBank mRNA | AB032165.1 | - | 2 - 3 | 193 | 79 | |
| AK142330.1 | - | 3 - 4 | 376 | 79 | ||
| AK154826.1 | - | 3 - 4 | 493 | 79 | ||
| AK160573.1 | - | 4 - 5 | 397 | 79 | ||
| BC018217.1 | - | 3 - 4 | 359 | 79 | ||
| BC036972.1 | - | 3 - 4 | 418 | 79 | ||
| BC066188.1 | - | 3 - 4 | 301 | 30476111 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC036972.1 transcripts.
Set Membership: |
|

Sfmbt1
Genomic Map