Assay Details
| Gene Symbol: Slc13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019481.2 | NP_062354.2 | 4 - 5 | 579 | 115 | |
| GenBank mRNA | AF199365.1 | - | 4 - 5 | 579 | 115 | |
| AF199366.1 | - | 4 - 5 | 579 | 115 | ||
| AK128899.1 | - | 4 - 5 | 579 | 115 | ||
| BC040789.1 | - | 4 - 5 | 553 | 115 | ||
| BC049981.1 | - | 4 - 5 | 561 | 5065026 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a1
Genomic Map