Assay Details
| Gene Symbol: Mbtps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167910.1 | NP_001161382.1 | 5 - 6 | 1181 | 75 | |
| NM_019709.4 | NP_062683.3 | 6 - 7 | 1330 | 75 | ||
| GenBank mRNA | AF094820.1 | - | 4 - 5 | 749 | 75 | |
| AK029048.1 | - | 5 - 6 | 1158 | 75 | ||
| AK122202.1 | - | 5 - 6 | 1147 | 75 | ||
| AK147336.1 | - | 5 - 6 | 1126 | 75 | ||
| AK147495.1 | - | 5 - 6 | 1155 | 75 | ||
| BC011533.1 | - | 5 - 6 | 1147 | 75 | ||
| BC054837.1 | - | 4 - 5 | 949 | 6490394 | 75 | |
| BC057198.1 | - | 6 - 7 | 1296 | 5310414 | 75 | |
| DQ913734.1 | - | 4 - 5 | 812 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbtps1
Genomic Map