Assay Details
| Gene Symbol: Cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021886.1 | NP_068686.1 | 2 - 3 | 264 | 114 | |
| GenBank mRNA | AB017634.1 | - | 2 - 3 | 264 | 114 | |
| AK006208.1 | - | 2 - 3 | 248 | 113 | ||
| AK011264.1 | - | 2 - 3 | 249 | 114 | ||
| AK011738.1 | - | 2 - 3 | 249 | 114 | ||
| BC025084.1 | - | 2 - 3 | 215 | 3501244 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenph
Genomic Map