Assay Details
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022879.2 | NP_075017.2 | 6 - 7 | 451 | 66 | |
| GenBank mRNA | AJ348866.1 | - | 5 - 6 | 361 | 66 | |
| AK146716.1 | - | 6 - 7 | 452 | 66 | ||
| BC061100.1 | - | 6 - 7 | 447 | 30300846 | 66 | |
| CA461005.1 | - | 6 - 7 | 488 | 66 | ||
| S70785.1 | - | 5 - 6 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10195 | S70785.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10195 | NM_022879.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10195 | AJ348866.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10195 | AK146716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P10195 | BC061100.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853783 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104189 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10195 | S70785.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10195 | NM_022879.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10195 | AJ348866.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10195 | AK146716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P10195 | BC061100.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246345 | NM_022879.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246345 | AJ348866.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565539 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104189 | - | - |
| Whole Mouse Genome G4122A | A_51_P246345 | NM_022879.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246345 | AJ348866.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101504 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116792 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104189 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103887 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103897 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103874 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09103873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09115704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09239671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09485801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09485802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09115702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09103897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09369616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09078104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565535 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384050 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449071_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384044 | - | - |
| Mouse Genome 430 2.0 Array | 1449071_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449071_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17898_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17898_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100020961.mm.1 | NM_022879.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020961.mm.1 | NM_022879.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020961.mm.1 | TC1100002156.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449071_at | - | - |
| Mouse Expression Set 430 | 1449071_PM_at | - | - |
| Murine Genome U74v2 Set | 100403_at | - | - |
| Murine Genome U74v2 Set | 100403_PM_at | - | - |
| Murine Genome U74v2 Set | 170292_at | - | - |
| Murine Genome U74v2 Set | 170292_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449071_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449071_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449071_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449071_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 519196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2480541 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480541 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2768253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2768253 | - | - |
| mouse-6 v1.1 (Illumina) | scl017898.5_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480541 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017898.5_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480541 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2768252 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2768252 | - | - |

Myl7
Genomic Map