Assay Details
| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025317.2 | NP_079593.1 | 1 - 2 | 153 | 146 | |
| GenBank mRNA | AK002348.1 | - | 1 - 2 | 155 | 146 | |
| AK003225.1 | - | 1 - 2 | 154 | 146 | ||
| AK009657.1 | - | 1 - 2 | 136 | 146 | ||
| BC056985.1 | - | 1 - 2 | 136 | 6818054 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288458 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174701 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105814 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105814 | - | - |
| Whole Mouse Genome G4122A | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | AK009657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | BC056985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | AK002348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | AK003225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372312 | NM_025317.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101654 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105814 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08624227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08624225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08672996 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08673000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08564435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08624243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288459 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27288457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08470928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08877091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288455 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10371109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10371108 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448699_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456788_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10371107 | - | - |
| Mouse Genome 430 2.0 Array | 1448699_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448699_at | - | - |
| Mouse Genome 430 2.0 Array | 1456788_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456788_at | - | - |
| Mouse Genome 430 2.0 Array | 1457378_at | - | - |
| Mouse Genome 430 2.0 Array | 1457378_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66047_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66047_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000018955.mm.1 | NM_025317.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018955.mm.1 | NM_025317.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018955.mm.1 | TC1000002534.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000018956.mm.1 | NM_025317.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018956.mm.1 | NM_025317.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000018956.mm.1 | TC1000002534.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448699_PM_at | - | - |
| Mouse Expression Set 430 | 1448699_at | - | - |
| Mouse Expression Set 430 | 1456788_PM_at | - | - |
| Mouse Expression Set 430 | 1456788_at | - | - |
| Murine Genome U74v2 Set | 99630_PM_at | - | - |
| Murine Genome U74v2 Set | 99630_at | - | - |
| Murine Genome U74v2 Set | 168866_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168866_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448699_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448699_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448699_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448699_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456788_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456788_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 343526 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 870403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 870403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000870403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755519 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755519 | - | - |
| mouse-6 v1.1 (Illumina) | scl37704.3.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 870403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37704.3.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755519 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755519 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37704.3.1_30-S | - | - |

Mrpl54
Genomic Map