Assay Details
| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025656.4 | NP_079932.2 | 2 - 3 | 303 | 90 | |
| GenBank mRNA | AK005928.1 | - | 2 - 3 | 305 | 90 | |
| AK007515.1 | - | 2 - 3 | 304 | 90 | ||
| AK013414.1 | - | 2 - 3 | 260 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gemin2
Genomic Map