Assay Details
| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025961.5 | NP_080237.1 | 4 - 5 | 735 | 70 | |
| GenBank mRNA | AK007325.1 | - | 4 - 5 | 735 | 70 | |
| AK145862.1 | - | 4 - 5 | 737 | 70 | ||
| AK145914.1 | - | 4 - 5 | 737 | 70 | ||
| AK146052.1 | - | 4 - 5 | 734 | 70 | ||
| AK146111.1 | - | 4 - 5 | 736 | 70 | ||
| AK151313.1 | - | 4 - 5 | 734 | 70 | ||
| AK152069.1 | - | 4 - 5 | 736 | 70 | ||
| AK153450.1 | - | 4 - 5 | 720 | 70 | ||
| AY625267.1 | - | 2 - 3 | 318 | 70 | ||
| BC003879.1 | - | 4 - 5 | 740 | 3495871 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatm
Genomic Map