Assay Details
| Gene Symbol: Cd82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136055.2 | NP_001129527.1 | 7 - 8 | 573 | 76 | |
| NM_001271430.1 | NP_001258359.1 | 6 - 7 | 629 | 76 | ||
| NM_001271431.1 | NP_001258360.1 | 6 - 7 | 497 | 76 | ||
| NM_001271432.1 | NP_001258361.1 | 8 - 9 | 792 | 76 | ||
| NM_001271461.1 | NP_001258390.1 | 7 - 8 | 584 | 76 | ||
| NM_001271462.1 | NP_001258391.1 | 7 - 8 | 596 | 76 | ||
| NM_007656.5 | NP_031682.1 | 7 - 8 | 705 | 76 | ||
| GenBank mRNA | AK135381.1 | - | 7 - 8 | 569 | 76 | |
| AK146912.1 | - | 7 - 8 | 569 | 76 | ||
| AK152871.1 | - | 7 - 8 | 700 | 76 | ||
| AK153070.1 | - | 7 - 8 | 700 | 76 | ||
| AK160907.1 | - | 6 - 7 | 551 | 76 | ||
| AK168097.1 | - | 7 - 8 | 656 | 76 | ||
| AK168515.1 | - | 7 - 8 | 675 | 76 | ||
| BC132559.1 | - | 7 - 8 | 675 | 40130836 | 76 | |
| BC132565.1 | - | 7 - 8 | 675 | 40130842 | 76 | |
| D14883.1 | - | 6 - 7 | 538 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd82
Genomic Map